Skip to Content
Merck
All Photos(1)

Key Documents

284548

Sigma-Aldrich

3,4-(Methylenedioxy)toluene

97%

Synonym(s):

5-Methyl-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

199-200 °C (lit.)

density

1.135 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc2OCOc2c1

InChI

1S/C8H8O2/c1-6-2-3-7-8(4-6)10-5-9-7/h2-4H,5H2,1H3

InChI key

GHPODDMCSOYWNE-UHFFFAOYSA-N

Application

3,4-(Methylenedioxy)toluene can be used as a cation scavenger for the selective cleavage of 4-(3,4-dimethoxyphenyl)benzyl (DMPBn) ethers affording the corresponding deprotected products in the presence of trifluoroacetic acid (TFA) in anhydrous CH2Cl2.
It can also be used as a starting material to prepare:
  • 1,3-Benzodioxole-5-carboxaldehyde via photooxidation using CBr4.
  • 6-Methyl-1,3-benzodioxole-5-carboxaldehyde by treating with dichloromethylmethyl ether and TiCl4.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visible light photocatalysis with CBr 4: a highly selective aerobic photooxidation of methylarenes to aldehydes
Shubhangi T, et al.
Royal Society of Chemistry Advances, 6(18), 14547-14551 (2016)
Synthesis of cicerfuran, an antifungal benzofuran, and some related analogues
Aslam SN, et al.
Tetrahedron, 62(17), 4214-4226 (2006)
Gregory M Rankin et al.
The Journal of organic chemistry, 78(11), 5264-5272 (2013-05-18)
A reliable reagent system for the cleavage of 4-(3,4-dimethoxyphenyl)benzyl (DMPBn) ethers under acidic conditions has been established. Treatment of DMPBn-protected mono- and pseudodisaccharides with TFA in anhydrous CH2Cl2 and 3,4-(methylenedioxy)toluene as a cation scavenger resulted in the selective cleavage of
The Dimethoxyphenylbenzyl Protecting Group: An Alternative to the p-Methoxybenzyl Group for Protection of Carbohydrates
Rankin GM, et al.
The Journal of Organic Chemistry, 78(11), 5264-5272 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service