Skip to Content
Merck
All Photos(3)

Key Documents

284432

Sigma-Aldrich

Octacosanoic acid

synthetic, ≥98%

Synonym(s):

Montanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)26CO2H
CAS Number:
Molecular Weight:
424.74
Beilstein:
1801616
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

91-93 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear, colorless

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C28H56O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28(29)30/h2-27H2,1H3,(H,29,30)

InChI key

UTOPWMOLSKOLTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octacosanoic acid is one of the components of the fruit of Hippophae rhamnoides. It is also present in the aerial part of Curcuma wenyujin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zheng-Ming Tao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 32(24), 2604-2606 (2008-03-15)
To investigate the chemical constituents from aerial part of Curcuma wenyujin. Compounds were isolated by repeated column chromatography on silica gel. Their structures were elucidated on the basis of spectral analysis and comparison with literature data. Six compounds were isolated
Effect of D-003 on a subconvulsive dose of kainic Acid in rats.
Daisy Carbajal et al.
Drugs in R&D, 5(6), 331-336 (2004-11-26)
Rui-Xia Zheng et al.
Natural product research, 23(15), 1451-1456 (2009-10-08)
Ten compounds were isolated from the fruits of Hippophae rhamnoides. On the basis of spectroscopic and chemical methods, the structures of these compounds were elucidated as hippophae cerebroside (1), oleanolic acid (2), ursolic acid (3), 19-alpha-hydroxyursolic acid (4), dulcioic acid
[Simple quantitative method of direct transesterification of higher fatty acid in biological samples].
V Z Lankin et al.
Voprosy meditsinskoi khimii, 17(3), 331-335 (1971-05-01)
R Menéndez et al.
Pharmacological research, 44(4), 299-304 (2001-10-11)
The present study was undertaken to investigate the effects of D003, a mixture of very long chain saturated fatty acids isolated and purified from sugar cane wax, on cholesterol biosynthesis in cultured fibroblasts. Cholesterol biosynthesis is regulated through feedback regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service