Skip to Content
Merck
All Photos(3)

Key Documents

282960

Sigma-Aldrich

Quinhydrone

97%

Synonym(s):

Hydroquinone: benzoquinone 1:1 complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)2 · C6H4O2
CAS Number:
Molecular Weight:
218.21
Beilstein:
3919222
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

composition

Carbon, 63.7-68.4%

mp

167-172 °C (lit.)

functional group

ketone

SMILES string

Oc1ccc(O)cc1.O=C2C=CC(=O)C=C2

InChI

1S/C6H6O2.C6H4O2/c2*7-5-1-2-6(8)4-3-5/h1-4,7-8H;1-4H

InChI key

BDJXVNRFAQSMAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinhydrone/methanol treatment for the measurement of carrier lifetime in crystalline silicon substrates has been reported. Surface passivation of silicon substrates by quinhydrone/ethanol treatment has been investigated.

Application

Quinhydrone is a general reagent used in potentiometric titrations. It can also be used as a π-acceptor in the formation of charge-transfer complex of analytes for spectrophotometric analysis.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface passivation effect of silicon substrates due to quinhydrone/ethanol treatment.
Takato H, et al.
Jpn J. App. Phys., Part I, 40(10A), L1003-L1003 (2001)
Photoinduced electron transfer from porphyrin to C60 in a C60| porphyrin double-layer photoelectrochemical cell.
Takahashi K, et al.
Journal of Electroanalytical Chemistry, 426(1-2), 85-90 (1997)
p-Benzosemiquinone radical anion on silver nanoparticles in water.
Tripathi G N R
Journal of the American Chemical Society, 125(5), 1178-1179 (2003)
Quinhydrone/methanol treatment for the measurement of carrier lifetime in silicon substrates.
Takato H, et al.
Jpn J. App. Phys., Part I, 41(8A), L870-L870 (2002)
F M Attia
Farmaco (Societa chimica italiana : 1989), 55(11-12), 659-664 (2001-02-24)
Three simple and selective methods are proposed for the determination of nortriptyline hydrochloride in bulk form and in tablets. The first two methods are based on the formation of charge-transfer complexes between the drug base as a n-donor and quinhydrone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service