Skip to Content
Merck
All Photos(2)

Key Documents

265322

Sigma-Aldrich

2,3-Difluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3OH
CAS Number:
Molecular Weight:
130.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

bp

54 °C/25 mmHg (lit.)

mp

39-42 °C (lit.)

functional group

fluoro

SMILES string

Oc1cccc(F)c1F

InChI

1S/C6H4F2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H

InChI key

RPEPGIOVXBBUMJ-UHFFFAOYSA-N

Application

2,3-Difluorophenol was used in the synthesis of material-poly{methyl[3-(2-hydroxy-3,4-difluoro)phenyl]propyl siloxane} (PMDFPS) sensitive to toxic organophosphate[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei He et al.
Talanta, 76(3), 698-702 (2008-07-01)
A new material-poly{methyl[3-(2-hydroxy-3,4-difluoro)phenyl]propyl siloxane}(PMDFPS) sensitive to toxic organophosphate vapor was synthesized with 2,3-difluorophenol, allyl bromide and poly (methyl hydrosiloxane) as raw materials, via O-alkylation, Claisen rearrange reaction and hydrosilylation reaction. This novel material was then coated on a quartz crystal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service