Skip to Content
Merck
All Photos(1)

Key Documents

264989

Sigma-Aldrich

2-Methyl-3-thiosemicarbazide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCSN(CH3)NH2
CAS Number:
Molecular Weight:
105.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
97%

Quality Level

Assay

97%

form

powder

mp

173-175 °C (lit.)

SMILES string

CN(N)C(N)=S

InChI

1S/C2H7N3S/c1-5(4)2(3)6/h4H2,1H3,(H2,3,6)

InChI key

IIGQLQZSWDUOBI-UHFFFAOYSA-N

General description

The rate constant for dihydroxy intermediate formation from 2-methyl-3-thiosemicarbazide was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of new amino acyl-4-thiazolidones.
Leite ACL, et al.
Quimica nova, 30(2), 284-286 (2007)
Reaction between various benzaldehydes and phenylhydroxylamine: special behaviour compared with other amines.
Brighente IM, et al.
J. Chem. Soc. Perkin Trans. II, 11, 1861-1864 (1991)
Franco Bisceglie et al.
Metallomics : integrated biometal science, 8(12), 1255-1265 (2016-11-15)
A comparative study between two bisthiosemicarbazones, 2,3-butanedione bis(4,4-dimethyl-3-thiosemicarbazone) and 2,3-butanedione bis(2-methyl-3-thiosemicarbazone), and their copper(ii) complexes is reported. The four compounds have been tested on a leukemia cell line U937 (p53-null) and on an adenocarcinoma cell line A549. The study includes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service