Skip to Content
Merck
All Photos(2)

Key Documents

252867

Sigma-Aldrich

2-(Benzyloxy)ethanol

98%

Synonym(s):

Ethylene glycol monobenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH2OH
CAS Number:
Molecular Weight:
152.19
Beilstein:
2043566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.521 (lit.)

bp

265 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

SMILES string

OCCOCc1ccccc1

InChI

1S/C9H12O2/c10-6-7-11-8-9-4-2-1-3-5-9/h1-5,10H,6-8H2

InChI key

CUZKCNWZBXLAJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Benzyloxy)ethanol has been used in:
  • 9-(2-hydroxyethyl)-linked synthesis of 2,6-diaminopurines
  • synthesis of benzyloxy ethyl methacrylate monomer by esterification with methacryloyl chloride
  • a base-free iridium-catalyzed direct alkylation of active methylene compounds with alcohols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A structure-based library approach to kinase inhibitors.
Norman TC, et al.
Journal of the American Chemical Society, 118(31), 7430-7431 (1996)
Poly (benzyloxy ethyl methacrylate): preparation by free radical polymerization and by group transfer polymerization; with subsequent debenzylation to form poly (2-hydroxyethyl methacrylate).
Dyakonov TA, et al.
Eur. Polymer J., 36(9), 1779-1793 (2000)
Masao Morita et al.
Chemical communications (Cambridge, England), (27), 2850-2852 (2007-07-05)
Base-free catalytic alpha-alkylation of active methylene compounds with primary alcohols was successfully achieved using an [IrCl(cod)](2) complex in the presence of PPh(3) to afford the corresponding saturated alpha-alkylated products in good yields.
Ugo Sidoli et al.
International journal of molecular sciences, 20(24) (2019-12-19)
During the last few decades, wet adhesives have been developed for applications in various fields. Nonetheless, key questions such as the most suitable polymer architecture as well as the most suitable chemical composition remain open. In this article, we investigate
Tsunehisa Hirose et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-05)
The retention behavior of a wide variety of stationary phases was compared in supercritical fluid chromatography (SFC) and normal-phase high-performance liquid chromatography (NP-HPLC). We also attempted to elucidate the retention behavior in SFC by investigating the selectivity of the different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service