Skip to Content
Merck
All Photos(1)

Key Documents

246751

Sigma-Aldrich

Polyethylenimine on silica gel

powder, 40-200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

particle size

40-200 mesh

SMILES string

O[SiH3].C1CN1

InChI

1S/C2H5N.H4OSi/c1-2-3-1;1-2/h3H,1-2H2;1H,2H3

InChI key

BPDVQMZCNJKUAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ion-exchange resin

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service