Skip to Content
Merck
All Photos(1)

Key Documents

239240

Sigma-Aldrich

Cesium

ingot, ≥99.95% trace metals basis

Synonym(s):

Caesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs
CAS Number:
Molecular Weight:
132.91
EC Number:
MDL number:
UNSPSC Code:
12141801
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 279 °C)

Quality Level

Assay

≥99.95% trace metals basis

form

ingot

reaction suitability

reagent type: reductant

packaging

pkg of Packaged in: Breakseal Ampule

resistivity

19 μΩ-cm, 0°C

bp

705 °C (lit.)

mp

28.5 °C (lit.)

density

1.873 g/mL at 25 °C (lit.)

SMILES string

[Cs]

InChI

1S/Cs

InChI key

TVFDJXOCXUVLDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cesium(Cs) is a highly reactive alkali metal known for its unique properties its uniqueproperties, such as a low melting point and high atomic mass, which enhance itsexceptional conductivity and reactivity. Its compounds, particularly cesiumlead halide perovskites, are extensively utilized in next-generation solarcells to achieve high efficiency and stability. Additionally, cesium isutilized in electron emission devices and photonic technologies.

Application

  • Cesium-containing triple cation perovskite solar cells: improved stability, reproducibility and high efficiency: Demonstrates how adding cesium to perovskite solar cells improves their stability and efficiency, making a significant advancement in solar technology (M Saliba et al., 2016).
  • Exotic stable cesium polynitrides at high pressure: Explores the formation of new cesium polynitrides under high pressure, indicating potential applications in high-energy-density materials (F Peng et al., 2015).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
Chemical communications (Cambridge, England), 49(3), 306-308 (2012-11-28)
A noncentrosymmetric chiral Strandberg-type polyoxometalate based material, Cs(4)Mo(5)P(2)O(22), that contains unusual one dimensional chains constructed from [Mo(5)P(2)O(23)](6-) units has been reported. Cs(4)Mo(5)P(2)O(22) shows a moderate SHG response, and is phase-matchable.
Dahu Ding et al.
Water research, 47(7), 2563-2571 (2013-03-14)
A novel biosorbent derived from agricultural residue - walnut shell (WS) is reported to remove cesium from aqueous solution. Nickel hexacyanoferrate (NiHCF) was incorporated into this biosorbent, serving as a high selectivity trap agent for cesium. Field emission scanning electron
Jun Koarashi et al.
Scientific reports, 2, 1005-1005 (2012-12-21)
The fate of (137)Cs derived from the Fukushima nuclear accident fallout and associated radiological hazards are largely dependent on its mobility in the surface soils of forest ecosystems. Thus, we quantified microbial and adsorptive retentions of (137)Cs in forest surface
Mao-sheng Miao
Nature chemistry, 5(10), 846-852 (2013-09-24)
The periodicity of the elements and the non-reactivity of the inner-shell electrons are two related principles of chemistry, rooted in the atomic shell structure. Within compounds, Group I elements, for example, invariably assume the +1 oxidation state, and their chemical
Min Lu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(6), 2099-2104 (2013-01-24)
Multidrug transporters belonging to the multidrug and toxic compound extrusion (MATE) family expel dissimilar lipophilic and cationic drugs across cell membranes by dissipating a preexisting Na(+) or H(+) gradient. Despite its clinical relevance, the transport mechanism of MATE proteins remains

Questions

1–2 of 2 Questions  
  1. What are the physical dimensions of the ampule that your 1 gram of cesium comes in? Thanks

    1 answer
    1. The dimensions of the glass ampoule are a follows:
      Diameter: 11.8 mm
      Height: 75 mm
      Nominal Volume: 2 mL

      Helpful?

  2. Hi, what are the dimensions of the glass tube that holds this 5g Cesium? Product No. 239240

    1 answer
    1. This product is packaged in breakseal ampules. See below for the ampule dimensions for each package size.
      239249-5G
      Total length: 13.3 cm
      Length of the closed portion containing the cesium: 6.3 cm
      Length of the arm: 7 cm
      Diameter of the ampoule: 11 mm ID, 15 mm OD


      239240-1G
      Total length: 13cm
      Length of the closed portion containing the cesium: 5 cm
      Length of the arm: 8 cm
      Diameter of the ampoule: 9-10 mm ID, 12 mm OD

      See the image below for a drawing of these ampules.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service