All Photos(2)
About This Item
Linear Formula:
C2H5C6H4COCH3
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.5293 (lit.)
bp
125 °C/20 mmHg (lit.)
mp
−20.6 °C (lit.)
density
0.993 g/mL at 25 °C (lit.)
functional group
ketone
SMILES string
CCc1ccc(cc1)C(C)=O
InChI
1S/C10H12O/c1-3-9-4-6-10(7-5-9)8(2)11/h4-7H,3H2,1-2H3
InChI key
NODGRWCMFMEGJH-UHFFFAOYSA-N
General description
4-Ethylacetophenone is one of the attractive components of a carob-peanut extract that elicited a positive behavioral response from Oryzaephilus surinamensis, S. granarius and C. ferrugineus.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
195.8 °F - closed cup
Flash Point(C)
91 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service