All Photos(1)
About This Item
Linear Formula:
BrC6H4CH2NH2 · HCl
CAS Number:
Molecular Weight:
222.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
mp
227-230 °C (lit.)
solubility
methanol: soluble 25 mg/mL, clear, colorless to yellow
functional group
amine
bromo
SMILES string
Cl.NCc1ccccc1Br
InChI
1S/C7H8BrN.ClH/c8-7-4-2-1-3-6(7)5-9;/h1-4H,5,9H2;1H
InChI key
MDEJOHXOXKDGOU-UHFFFAOYSA-N
General description
2-Bromobenzylamine hydrochloride reacts with sulfonyl chloride to give sulfonamide.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A tandem radical approach to the ABCE-rings of the Aspidosperma and Strychnos alkaloids.
Hilton ST, et al.
Chemical Communications (Cambridge, England), 2, 209-210 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service