Skip to Content
Merck
All Photos(2)

Key Documents

224081

Sigma-Aldrich

Chlorohydroquinone

technical grade, 85%

Synonym(s):

2-Chloro-1,4-dihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
144.56
Beilstein:
636835
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

85%

bp

263 °C (lit.)

mp

100-104 °C (lit.)

SMILES string

Oc1ccc(O)c(Cl)c1

InChI

1S/C6H5ClO2/c7-5-3-4(8)1-2-6(5)9/h1-3,8-9H

InChI key

AJPXTSMULZANCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Libis et al.
ACS synthetic biology, 5(10), 1076-1085 (2016-10-22)
Detection of chemical signals is critical for cells in nature as well as in synthetic biology, where they serve as inputs for designer circuits. Important progress has been made in the design of signal processing circuits triggering complex biological behaviors
Carolyne Caetano Gonçalves et al.
Frontiers in microbiology, 11, 1081-1081 (2020-06-26)
Lignin is an abundant cell wall component, and it has been used mainly for generating steam and electricity. Nevertheless, lignin valorization, i.e. the conversion of lignin into high value-added fuels, chemicals, or materials, is crucial for the full implementation of
Z Mehmood et al.
Chemosphere, 34(11), 2281-2291 (1997-06-01)
The metabolism of the environmental pollutant and hepatocarcinogen 2,4-dichlorophenol (2,4-DCP) was studied using microsomal fractions and whole-cells of Saccharomyces cerevisiae containing human cytochrome P450 3A4. 2,4-DCP exhibited a typical type I substrate binding spectrum with a K, of 75 microM.
Guadalupe Albarrán et al.
Environmental science and pollution research international, 26(17), 17055-17065 (2019-04-19)
Radiolytic oxidation of 2,4-dichlorophenol (2,4-DClP) in aqueous solutions demonstrated that ·OH predominantly adds to the unsubstituted positions of the aromatic ring and that elimination of chloride at the 4 position is important because the -OH group enhances the electron density
Guadalupe Albarrán et al.
Environmental science and pollution research international, 27(18), 22855-22864 (2020-04-24)
Radiation-induced degradation of chlorobenzene was conducted at 0.1, 0.4, 0.5, 0.7, and 1.0 mmol/dm3 concentrations in aerated environment and at 1.0 mmol/dm3 in oxygen-free and N2O-saturated solutions. The results demonstrated that the elimination of chloride is important when the solution is oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service