Skip to Content
Merck
All Photos(4)

Documents

215554

Sigma-Aldrich

Copper(I) iodide

99.999% trace metals basis

Synonym(s):

Cuprous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuI
CAS Number:
Molecular Weight:
190.45
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150105
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

10 mmHg ( 656 °C)

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

bp

1290 °C/1 atm (decomp)(lit.)

mp

605 °C (lit.)

solubility

dilute aqueous acid: insoluble(lit.)

density

5.62 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu+].[I-]

InChI

1S/Cu.HI/h;1H/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Copper(I) iodide can be used as a catalyst for:
  • For arylation of pyrazole and derivatives by coupling arenediazonium species under mild and ligand-free conditions.
  • Selective cross-coupling of dibromo alkenes and heteronucleophiles which leads to numerous building blocks such as 1-bromoenamides, ynamides, ketene N, N-acetals, 1-bromoenol ethers, ynol ethers, ketene O, O-acetals, and vinyl phosphonates.
  • Stereospecific and the regioselective reaction of silacyclopropanes with carbonyl compounds.
It can also be used as a starting material in the preparation of diphenylphosphinepyridine-derivatives (PyrPHOS)-copper iodide complexes which are suitable for use in OLEDs.

Features and Benefits

  • Low toxicity
  • Mild reaction condition
  • High purity ensures reproducibility and avoids cocatalysis by contaminants.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuangzhuang Ma et al.
Advanced materials (Deerfield Beach, Fla.), 33(2), e2001367-e2001367 (2020-11-24)
White light-emitting diodes (WLEDs) are promising next-generation solid-state light sources. However, the commercialization route for WLED production suffers from challenges in terms of insufficient color-rendering index (CRI), color instability, and incorporation of rare-earth elements. Herein, a new two-component strategy is
Synlett, 1836-1836 (2007)
Synlett, 1917-1917 (2007)
Roberta Berrino et al.
The Journal of organic chemistry, 77(5), 2537-2542 (2012-01-31)
Free NH 3,3-diarylacrylamides are cyclized to substituted 2-quinolones in the presence of CuI, PPh(3), and KO-t-Bu in o-xylene at 100 °C. The reaction proceeds through a C-H functionalization/C-N bond formation process. With unsymmetrical 3,3-diarylacrylamides, high selectivity is observed using substrates
Min Jiang et al.
Organic letters, 14(6), 1420-1423 (2012-03-08)
A domino synthesis of 5,12-dihydroindolo[2,1-b]quinazoline derivatives via copper-catalyzed Ullmann-type intermolecular C-C and intramolecular C-N couplings is reported. Good yields of various 5,12-dihydroindolo[2,1-b]quinazoline derivatives were obtained. Reaction scopes, limitations, and the reaction mechanism are discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service