Skip to Content
Merck
All Photos(2)

Key Documents

212814

Sigma-Aldrich

1-Naphthol-3,6-disulfonic acid disodium salt hydrate

technical grade

Synonym(s):

4-Hydroxy-2,7-naphthalenedisulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H5(SO3Na)2 · xH2O
CAS Number:
Molecular Weight:
348.26 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

impurities

10% sodium chloride

SMILES string

O.[Na+].[Na+].Oc1cc(cc2cc(ccc12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C10H8O7S2.2Na.H2O/c11-10-5-8(19(15,16)17)4-6-3-7(18(12,13)14)1-2-9(6)10;;;/h1-5,11H,(H,12,13,14)(H,15,16,17);;;1H2/q;2*+1;/p-2

InChI key

BJNIHPYLVMUUPO-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile Synthesis of Novel 1-(2, 4-Diaminephenoxy)-3, 6-naphthalenedisulfonic Acid.
DU W-S, et al.
Fine Chemicals / ????, 2(2011), 023-023 (2011)
Mirabelle Prémont-Schwarz et al.
The journal of physical chemistry. B, 117(16), 4594-4603 (2013-01-12)
The 1-naphthol molecule has been the subject of intense research activity for the past 60 years due to its complex behavior as a photoacid upon optical excitation. We have utilized femtosecond mid-infrared spectroscopy and time-resolved fluorescence spectroscopy to investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service