Skip to Content
Merck
All Photos(1)

Documents

17640

Sigma-Aldrich

1-Bromonaphthalene

≥95%

Synonym(s):

1-Naphthyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Br
CAS Number:
Molecular Weight:
207.07
Beilstein:
1906414
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39060108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

color

slightly yellow to deep brown-yellow

refractive index

n20/D 1.6570 (lit.)

bp

133-134 °C/10 mmHg (lit.)

mp

−2-−1 °C (lit.)

solubility

H2O: slightly soluble
alcohol: miscible
benzene: miscible
chloroform: miscible
diethyl ether: miscible

density

1.48 g/mL at 20 °C (lit.)

SMILES string

Brc1cccc2ccccc12

InChI

1S/C10H7Br/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

DLKQHBOKULLWDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The vibrational spectra of 1-bromonaphthalene has been studied.

Application

1-Bromonaphthalene was used as a solvent for the exfoliation and dispersion of hexabenzocoronene (HBC).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinzhen Du et al.
Journal of colloid and interface science, 274(2), 645-651 (2004-05-18)
The binding of polyethylene glycol (10) n-octylphenyl ether (OPE) and polyethylene glycol (10) tert-octylphenyl ether (Triton X-100, TX) to beta-cyclodextrin (beta-CD) and heptakis(2,3- beta-dimethyl)- beta-CD (DM- beta-CD) was described in detail by surface tension, steady-state fluorescence of OPE and TX
J Marguerite Hughes et al.
Journal of the American Chemical Society, 134(29), 12168-12179 (2012-06-22)
We have studied the exfoliation and dispersion of hexabenzocoronene (HBC) in 28 different solvents. We see a wide range of dispersed concentrations and aggregation states, all of which can be related to the solvent properties. To a first approximation, the
M Arivazhagan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 553-560 (2011-10-01)
In this work, the experimental and theoretical vibrational spectra of 1-bromonaphthalene (1-BN) were studied. FTIR and FT Raman spectra were recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. The structural and spectroscopic data of the molecule in the
Xin-zhen Du et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(4), 402-406 (2005-03-16)
Room-temperature phosphorescence of 1-BrN induced by a combination of OPE-10 and Triton X-100 with beta-CD was comparatively studied. In terms of molecular size and dimensions of beta-CD, the octyl group and phenyl group of OPE-10 and Triton X-100 were incorporated
M B Habash et al.
Microbiology (Reading, England), 143 ( Pt 8), 2569-2574 (1997-08-01)
The physico-chemical cell-surface properties of Pseudomonas aeruginosa AK1 and its adhesion to silicone rubber under flow were compared for cells suspended in phosphate-buffered saline (PBS) or PBS supplemented with 2% nutrient broth. Addition of 2% nutrient broth to cells suspended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service