15345
Boc-Glu-OH
≥98.0% (T)
Synonym(s):
Boc-L-glutamic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3COCONHCH(COOH)CH2CH2COOH
CAS Number:
Molecular Weight:
247.25
Beilstein:
2418563
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0% (T)
optical activity
[α]20/D −14.5±2°, c = 1% in methanol
reaction suitability
reaction type: Boc solid-phase peptide synthesis
mp
~110 °C (dec.)
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
CC(C)(C)OC(=O)N[C@@H](CCC(O)=O)C(O)=O
InChI
1S/C10H17NO6/c1-10(2,3)17-9(16)11-6(8(14)15)4-5-7(12)13/h6H,4-5H2,1-3H3,(H,11,16)(H,12,13)(H,14,15)/t6-/m0/s1
InChI key
AQTUACKQXJNHFQ-LURJTMIESA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shao-Fei Zhang et al.
Journal of biomedical materials research. Part A, 106(6), 1687-1696 (2018-02-23)
Thrombotic events affect many individuals in a number of ways, all of which can cause significant morbidity and mortality. Nattokinase (NK), as a novel thrombolytic drug, has been used for thrombolytic therapy. It not only possesses plasminogen activator activity, but
Mengjie Huang et al.
Journal of biomedical materials research. Part A, 107(8), 1824-1831 (2019-04-24)
With the frequent occurrence of thrombus diseases, thrombus has become a factor endangering human health. Nattokinase (NK) is a new generation of thrombolytic drug with efficient thrombolytic effect and no major side effects. However, it is easily inactivated in external
Jie Shi et al.
Chemical science, 8(9), 6188-6195 (2017-10-11)
As a sudden inflammation of the pancreas, acute pancreatitis presents severe complications and a high mortality rate, despite treatment. Lipase in serum serves as an essential biomarker of acute pancreatitis and even pancreatic cancer. Therefore, developing robust, convenient and sensitive
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service