Skip to Content
Merck
All Photos(1)

Key Documents

125504

Sigma-Aldrich

Cyclopropylamine

98%

Synonym(s):

Aminocyclopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5NH2
CAS Number:
Molecular Weight:
57.09
Beilstein:
741858
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

4.67 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

autoignition temp.

527 °F

refractive index

n20/D 1.420 (lit.)

bp

49-50 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

SMILES string

NC1CC1

InChI

1S/C3H7N/c4-3-1-2-3/h3H,1-2,4H2

InChI key

HTJDQJBWANPRPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclopropylamine(CPA) has been used in the synthesis of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It has been used in the synthesis of Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Biochem/physiol Actions

Cyclopropylamine inactivates cytochrome P450 enzymes by a mechanism involving initial one-electron oxidation at nitrogen followed by scission of the cyclopropane ring leading to covalent modification of the enzyme. It is a mechanism-based inhibitor of quinoprotein methylamine dehydrogenase from Paracoccus denitrificans.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and Antifungal Activity of 5-[2-(Alkylamino) pyrimidin-4-yl]-4-phenylthiazol-2-cycloalkylamine Derivatives on Phytophthora capsici.
Nam Sw, et al.
J. Korean Chem. Soc., 54(3), 395-402 (2011)
Coordination Mode vs. Anticancer Activity of the Platinum (II) Complexes Involving Sulfur-Containing Ylidenemalonate Ligands.
Sakai N, et al.
Bull. Korean Chem. Soc., 19(12), 1377-1379 (1998)
Dapeng Sun et al.
FEBS letters, 517(1-3), 172-174 (2002-06-14)
Cyclopropylamine is a mechanism-based inhibitor of the quinoprotein methylamine dehydrogenase (MADH) from Paracoccus denitrificans. The resulting inactivation is accompanied by the formation of a covalent cross-link between the alpha and beta subunits of MADH. The results of site-directed mutagenesis studies
Danny Gauvreau et al.
The Journal of organic chemistry, 75(12), 4078-4085 (2010-05-18)
The evolution of scalable, economically viable synthetic approaches to the potent and selective prostaglandin EP4 antagonist 1 is presented. The chromatography-free synthesis of multikilogram quantities of 1 using a seven-step sequence (six in the longest linear sequence) is described. This
Soda Chanthamath et al.
Organic letters, 15(4), 772-775 (2013-01-31)
The Ru(II)-Pheox-catalyzed asymmetric cyclopropanation of vinylcarbamates with diazoesters resulted in the corresponding cyclopropylamine derivatives in high yield and excellent diastereoselectivity (up to 96:4) and enantioselectivity (up to 99% ee).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service