Merck
  • Pseudo-Interphase of Liposome Promotes 1,3-Dipolar Cycloaddition Reaction of Benzonitrile Oxide and N-Ethylmaleimide in Aqueous Solution.
Pseudo-Interphase of Liposome Promotes 1,3-Dipolar Cycloaddition Reaction of Benzonitrile Oxide and N-Ethylmaleimide in Aqueous Solution.
The journal of physical chemistry. B (2015-07-07)
Fumihiko Iwasaki, Keishi Suga, Hiroshi Umakoshi
ABSTRACT
The hydrophobic interior of a liposome membrane was used as a platform for the organic synthesis of hydrophobic compounds in water. The 1,3-dipolar cycloaddition of benzonitrile oxide (BNO) and N-ethylmaleimide (EMI) in liposome suspensions was carried out, and an increase in the reaction rate constant was observed depending on the liposome characteristics. While the reaction rate constant in 1,4-dioxane was 1.5 times higher than that in water, the reaction rate constant in an aqueous solution of cationic 1,2-dioleoyl-3-trimethylammonium-propane (DOTAP) liposome was 3 times higher than in water. The amount of substrate, BNO, accumulated in the DOTAP liposome was higher than that in 1,2-dipalmitoyl-3-trimethylammonium-propane (DPTAP), indicating that BNO prefers to be distributed in the liposome membrane in the liquid-disordered phase. The membrane polarity, GP340, as monitored by Laurdan, varied with the presence of BNO, while EMI slightly affected the membrane properties of the liposomes. These results suggest that the pseudo-interphase afforded by the liposome membrane can promote the 1,3-dipolar cycloaddition between BNO and EMI in water.
Sigma-Aldrich
Sodium hypochlorite solution, CP
Sigma-Aldrich
Sodium hypochlorite solution, CP
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine, ≥97.0% (HPLC), suitable for fluorescence
Sigma-Aldrich
6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt