Merck
  • Social dominance and reproductive differentiation mediated by dopaminergic signaling in a queenless ant.
Social dominance and reproductive differentiation mediated by dopaminergic signaling in a queenless ant.
The Journal of experimental biology (2015-02-18)
Yasukazu Okada, Ken Sasaki, Satoshi Miyazaki, Hiroyuki Shimoji, Kazuki Tsuji, Toru Miura
ABSTRACT
In social Hymenoptera with no morphological caste, a dominant female becomes an egg layer, whereas subordinates become sterile helpers. The physiological mechanism that links dominance rank and fecundity is an essential part of the emergence of sterile females, which reflects the primitive phase of eusociality. Recent studies suggest that brain biogenic amines are correlated with the ranks in dominance hierarchy. However, the actual causality between aminergic systems and phenotype (i.e. fecundity and aggressiveness) is largely unknown due to the pleiotropic functions of amines (e.g. age-dependent polyethism) and the scarcity of manipulation experiments. To clarify the causality among dominance ranks, amine levels and phenotypes, we examined the dynamics of the aminergic system during the ontogeny of dominance hierarchy in the queenless ant Diacamma sp., which undergoes rapid physiological differentiation based on dominance interactions. Brain dopamine levels differed between dominants and subordinates at day 7 after eclosion, although they did not differ at day 1, reflecting fecundity but not aggressiveness. Topical applications of dopamine to the subordinate workers induced oocyte growth but did not induce aggressiveness, suggesting the gonadotropic effect of dopamine. Additionally, dopamine receptor transcripts (dopr1 and dopr2) were elevated in the gaster fat body of dominant females, suggesting that the fat body is a potential target of neurohormonal dopamine. Based on this evidence, we suggest that brain dopamine levels are elevated in dominants as a result of hierarchy formation, and differences in dopamine levels cause the reproductive differentiation, probably via stimulation of the fat body.
Sigma-Aldrich
Acetonitrile, JIS 5000
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, JIS 1000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, ≥98.0% (T), pellets
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid