574724
Acetonitrile solution
suitable for HPLC, contains 0.05 % (v/v) trifluoroacetic acid
Synonym(s):
ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide
About This Item
Recommended Products
Product Name
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
vapor density
1.41 (vs air)
Quality Level
vapor pressure
72.8 mmHg
form
liquid
contains
0.05 % (v/v) trifluoroacetic acid
expl. lim.
8 % (lit.)
technique(s)
HPLC: suitable
bp
81-82 °C (lit.)
mp
-48 °C (lit.)
density
0.782 g/mL at 25 °C
format
mixture
SMILES string
CC#N
InChI
1S/C2H3N/c1-2-3/h1H3
InChI key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Preparation Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
42.8 °F - closed cup
Flash Point(C)
6 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service