Merck
  • Nickel(II) complexes of pentadentate N5 ligands as catalysts for alkane hydroxylation by using m-CPBA as oxidant: a combined experimental and computational study.
Nickel(II) complexes of pentadentate N5 ligands as catalysts for alkane hydroxylation by using m-CPBA as oxidant: a combined experimental and computational study.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-08-08)
Muniyandi Sankaralingam, Mani Balamurugan, Mallayan Palaniandavar, Prabha Vadivelu, Cherumuttathu H Suresh
ABSTRACT
A new family of nickel(II) complexes of the type [Ni(L)(CH(3)CN)](BPh(4))(2), where L=N-methyl-N,N',N'-tris(pyrid-2-ylmethyl)-ethylenediamine (L1, 1), N-benzyl-N,N',N'-tris(pyrid-2-yl-methyl)-ethylenediamine (L2, 2), N-methyl-N,N'-bis(pyrid-2-ylmethyl)-N'-(6-methyl-pyrid-2-yl-methyl)-ethylenediamine (L3, 3), N-methyl-N,N'-bis(pyrid-2-ylmethyl)-N'-(quinolin-2-ylmethyl)-ethylenediamine (L4, 4), and N-methyl-N,N'-bis(pyrid-2-ylmethyl)-N'-imidazole-2-ylmethyl)-ethylenediamine (L5, 5), has been isolated and characterized by means of elemental analysis, mass spectrometry, UV/Vis spectroscopy, and electrochemistry. The single-crystal X-ray structure of [Ni(L(3))(CH(3)CN)](BPh(4))(2) reveals that the nickel(II) center is located in a distorted octahedral coordination geometry constituted by all the five nitrogen atoms of the pentadentate ligand and an acetonitrile molecule. In a dichloromethane/acetonitrile solvent mixture, all the complexes show ligand field bands in the visible region characteristic of an octahedral coordination geometry. They exhibit a one-electron oxidation corresponding to the Ni(II) /Ni(III) redox couple the potential of which depends upon the ligand donor functionalities. The new complexes catalyze the oxidation of cyclohexane in the presence of m-CPBA as oxidant up to a turnover number of 530 with good alcohol selectivity (A/K, 7.1-10.6, A=alcohol, K=ketone). Upon replacing the pyridylmethyl arm in [Ni(L1)(CH(3)CN)](BPh(4))(2) by the strongly σ-bonding but weakly π-bonding imidazolylmethyl arm as in [Ni(L5)(CH(3)CN)](BPh(4))(2) or the sterically demanding 6-methylpyridylmethyl ([Ni(L3)(CH(3)CN)](BPh(4))(2) and the quinolylmethyl arms ([Ni(L4)(CH(3)CN)](BPh(4))(2), both the catalytic activity and the selectivity decrease. DFT studies performed on cyclohexane oxidation by complexes 1 and 5 demonstrate the two spin-state reactivity for the high-spin [(N5)Ni(II)-O(.)] intermediate (ts1(hs), ts2(doublet)), which has a low-spin state located closely in energy to the high-spin state. The lower catalytic activity of complex 5 is mainly due to the formation of thermodynamically less accessible m-CPBA-coordinated precursor of [Ni(II) (L5)(OOCOC(6)H(4)Cl)](+) (5 a). Adamantane is oxidized to 1-adamantanol, 2-adamantanol, and 2-adamantanone (3°/2°, 10.6-11.5), and cumene is selectively oxidized to 2-phenyl-2-propanol. The incorporation of sterically hindering pyridylmethyl and quinolylmethyl donor ligands around the Ni(II) leads to a high 3°/2° bond selectivity for adamantane oxidation, which is in contrast to the lower cyclohexane oxidation activities of the complexes.
Sigma-Aldrich
Sodium triacetoxyborohydride, 97%
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
Sigma-Aldrich
Cyclohexane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethyl ether, ≥99.5%
Sigma-Aldrich
Cyclohexane, JIS special grade
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Cyclohexane, suitable for HPLC