Skip to Content
Merck
All Photos(1)

Documents

09-1810

Sigma-Aldrich

Diethyl ether

≥99.0%, SAJ first grade

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

product name

Diethyl ether, SAJ first grade, ≥99.0%

grade

SAJ first grade

vapor density

2.6 (vs air)

vapor pressure

28.5 psi

Assay

≥99.0%

form

liquid

autoignition temp.

320 °F

expl. lim.

36.5 %

availability

available only in Japan

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Determination of chloropropanol esters and glycidyl esters in nutritional foods by gas chromatography-tandem mass spectrometry based on acid hydrolysis and solid-phase extraction.: This study utilizes diethyl ether in the solid-phase extraction process to determine harmful esters in food products, showcasing its role in enhancing the efficiency of contaminant isolation and detection in complex matrices (Li Y et al., 2024).

  • Cloning, Expression, Characterization and Immobilization of a Recombinant Carboxylesterase from the Halophilic Archaeon, Halobacterium salinarum NCR-1.: Research employs diethyl ether for the extraction and purification steps in enzyme characterization, underlining its critical utility in biotechnological applications for enzyme stability and activity studies (Ortega-de la Rosa ND et al., 2024).

  • The Analysis of Leontopodium leontopodioides (Willd.) Beauv. Chemical Composition by GC/MS and UPLC-Q-Orbitrap MS.: Diethyl ether is used as an extraction solvent to analyze the chemical composition of medicinal plants, highlighting its effectiveness in capturing a broad spectrum of volatile and semi-volatile compounds (Chen Y et al., 2024).

  • Chemical composition and antimicrobial study of Crossobamon orientalis body oil.: In this research, diethyl ether is used for the extraction of essential oils, demonstrating its efficacy in obtaining bioactive compounds for antimicrobial studies (Ahmad S et al., 2024).

  • A Static Headspace Gas Chromatography Method for Quantitation of Residual Solvents in Nanoformulations.: Utilizes diethyl ether in a novel static headspace analysis technique for the quantification of residual solvents, showing its applicability in ensuring safety and compliance in pharmaceutical formulations (Kattel K et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Juan E Camacho Londoño et al.
European heart journal, 36(33), 2257-2266 (2015-06-13)
Pathological cardiac hypertrophy is a major predictor for the development of cardiac diseases. It is associated with chronic neurohumoral stimulation and with altered cardiac Ca(2+) signalling in cardiomyocytes. TRPC proteins form agonist-induced cation channels, but their functional role for Ca(2+)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Julia J Griese et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17189-17194 (2013-10-09)
Although metallocofactors are ubiquitous in enzyme catalysis, how metal binding specificity arises remains poorly understood, especially in the case of metals with similar primary ligand preferences such as manganese and iron. The biochemical selection of manganese over iron presents a
Alessandra Forni et al.
Journal of molecular graphics & modelling, 38, 31-39 (2012-10-23)
The solvent effect on the I⋯O halogen bonding in complexes of iodobenzene derivatives with formaldehyde has been investigated by systematically varying the substituents on the iodobenzene ring. Calculations have been performed at MP2 and DFT levels of theory, using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service