Merck
  • Integrated TE and TM optical circulators on ultra-low-loss silicon nitride platform.
Integrated TE and TM optical circulators on ultra-low-loss silicon nitride platform.
Optics express (2013-03-14)
Paolo Pintus, Fabrizio Di Pasquale, John E Bowers
ABSTRACT
In this paper, we present two four-port optical circulators for TE and TM modes, respectively. Exploiting the recent technological development concerning Ce:YIG pulse laser deposition on silicon nitride platform, we design two integrated circulators, which can be used to implement several functions in integrated optics, such as de-interleavers, input/output amplifier isolators and output laser isolators. The proposed devices combine the benefit of low loss silicon nitride waveguides with the non-reciprocal properties of magneto-optical materials. The ring cross-section has been optimized in order to maximize the non-reciprocal phase shift and finally the scattering coefficients have been computed using the transfer matrix method. The material stability and refractive index regularity of silicon nitride, the small micro-ring footprint, and the high wavelength selectivity make these devices particularly attractive.
Sigma-Aldrich
Silicon nitride, predominantly β-phase, ≤10micron primary particle size
Sigma-Aldrich
Silicon nitride, predominantly α-phase, ≤10 micron
Sigma-Aldrich
Silicon nitride, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Silicon nitride, nanopowder, <50 nm particle size (spherical), ≥98.5% trace metals basis