Merck
  • Clofibrate in combination with phototherapy for unconjugated neonatal hyperbilirubinaemia.
Clofibrate in combination with phototherapy for unconjugated neonatal hyperbilirubinaemia.
The Cochrane database of systematic reviews (2012-12-14)
Maryam Gholitabar, Hugh McGuire, Janet Rennie, Donal Manning, Rosalind Lai
ABSTRACT
There are many pathological conditions leading to an elevated unconjugated bilirubin level (hyperbilirubinaemia) in neonates. Currently the standard therapies for unconjugated hyperbilirubinaemia include phototherapy and exchange transfusion. In addition to phototherapy, clofibrate has been studied as a treatment for hyperbilirubinaemia in several countries. To determine the efficacy and safety of clofibrate in combination with phototherapy versus phototherapy alone in unconjugated neonatal hyperbilirubinaemia. Randomised controlled trials were identified by searching MEDLINE (1950 to April 2012) before being translated for use in The Cochrane Library, EMBASE 1980 to April 2012 and CINAHL databases. All searches were re-run on 2 April 2012. We included trials where neonates with hyperbilirubinaemia received either clofibrate in combination with phototherapy or phototherapy alone or placebo in combination with phototherapy. Data were extracted and analysed independently by two review authors (MG and HM). Treatment effects on the following outcomes were determined: mean change in bilirubin levels, mean duration of treatment with phototherapy, number of exchange transfusions needed, adverse effects of clofibrate, bilirubin encephalopathy and neonatal mortality. Study authors were contacted for additional information. Studies were analysed for methodological quality in a 'Risk of bias' table. Fifteen studies (two including preterm neonates and 13 including term neonates) were included in this review. All but one of the included studies were conducted in Iran. For preterm neonates, there was a significantly lower bilirubin level in the 100 mg/kg clofibrate group compared to the control group with a mean difference of -1.37 mg/dL (95% CI -2.19 mg/dL to -0.55 mg/dL) (-23 µmol/L; 95% CI -36 µmol/L to -9 µmol/L) after 48 hours. For the term neonates, there were significantly lower bilirubin levels in the clofibrate group compared to the control group after both 24 and 48 hours of treatment with a weighted mean difference of -2.14 mg/dL (95% CI -2.53 mg/dL to -1.75 mg/dL) (-37 µmol/L; 95% CI -43 µmol/L to -30 µmol/L] and -1.82 mg/dL (95% CI -2.25 mg/dL to -1.38 mg/dL) (-31 µmol/L; 95% CI -38 µmol/L to -24 µmol/L), respectively.There was a significantly lower duration of phototherapy in the clofibrate group compared to the control group for both preterm and term neonates with a weighted mean difference of -23.82 hours (95% CI -30.46 hours to -17.18 hours) and -25.40 hours (95% CI -28.94 hours to -21.86 hours), respectively.None of the studies reported on bilirubin encephalopathy rates, neonatal mortality rates, or the levels of parental or staff satisfactions with the interventions. There are insufficient data from different countries on the use of clofibrate in combination with phototherapy for hyperbilirubinaemia to make recommendations for practice. There is a need for larger trials to determine how effective clofibrate is in reducing the need for, and duration of, phototherapy in term and preterm infants with hyperbilirubinaemia.
Product Description
Supelco
Clofibrate, analytical standard
Sigma-Aldrich
Clofibrate, liquid
Clofibrate, European Pharmacopoeia (EP) Reference Standard