Merck
  • Aerobic oxidative amidation of aromatic and cinnamic aldehydes with secondary amines by CuI/2-pyridonate catalytic system.
Aerobic oxidative amidation of aromatic and cinnamic aldehydes with secondary amines by CuI/2-pyridonate catalytic system.
The Journal of organic chemistry (2012-09-26)
Mingwen Zhu, Ken-ichi Fujita, Ryohei Yamaguchi
ABSTRACT
A simple and convenient CuI/2-pyridonate catalytic system for the oxidative amidation of aldehydes with secondary amines has been developed. With this system, a variety of useful arylamides have been synthesized in moderate to good yields in the presence of small amount of copper catalyst and the pyridonate ligand, generating only water as a coproduct. Synthesis of cinnamamides was also achieved by the reactions of cinnamaldehydes with secondary amines in moderate yields. Air was successfully employed as a green oxidant in this catalytic system, achieving a safe and atom-efficient system for the synthesis of amides.
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
Copper(I) iodide, 98%