Skip to Content
Merck
All Photos(3)

Key Documents

205540

Sigma-Aldrich

Copper(I) iodide

98%

Synonym(s):

Cuprous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuI
CAS Number:
Molecular Weight:
190.45
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150105
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

10 mmHg ( 656 °C)

Assay

98%

form

powder

reaction suitability

reaction type: click chemistry
reagent type: catalyst
core: copper

mp

605 °C (lit.)

density

5.62 g/mL at 25 °C (lit.)

SMILES string

[Cu+].[I-]

InChI

1S/Cu.HI/h;1H/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P H Mee et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(11), 3294-3308 (2005-03-24)
The combination of copper(I) iodide and cesium fluoride significantly enhances the Stille reaction. After extensive optimisation, a variety of electronically unfavourable and sterically hindered substrates were coupled in very high yields under mild conditions.
The high-temperature structural behaviour of copper (I) iodide.
Keen DA and Hull S.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 7(29), 5793-5793 (1995)
Alkynyl sulfides and selenides from alkynyl bromides and diorganoyl chalcogenides promoted by copper (I) iodide.
Braga AL, et al.
Tetrahedron Letters, 34(3), 393-394 (1993)
Min Jiang et al.
Organic letters, 14(6), 1420-1423 (2012-03-08)
A domino synthesis of 5,12-dihydroindolo[2,1-b]quinazoline derivatives via copper-catalyzed Ullmann-type intermolecular C-C and intramolecular C-N couplings is reported. Good yields of various 5,12-dihydroindolo[2,1-b]quinazoline derivatives were obtained. Reaction scopes, limitations, and the reaction mechanism are discussed.
Mingwen Zhu et al.
The Journal of organic chemistry, 77(20), 9102-9109 (2012-09-26)
A simple and convenient CuI/2-pyridonate catalytic system for the oxidative amidation of aldehydes with secondary amines has been developed. With this system, a variety of useful arylamides have been synthesized in moderate to good yields in the presence of small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service