Merck
  • Hydroxocobalamin versus sodium thiosulfate for the treatment of acute cyanide toxicity in a swine (Sus scrofa) model.
Hydroxocobalamin versus sodium thiosulfate for the treatment of acute cyanide toxicity in a swine (Sus scrofa) model.
Annals of emergency medicine (2012-03-06)
Vikhyat S Bebarta, Rebecca L Pitotti, Patricia Dixon, Julio R Lairet, Anneke Bush, David A Tanen
ABSTRACT
We compare the efficacy of hydroxocobalamin to sodium thiosulfate to reverse the depressive effects on mean arterial pressure in a swine model of acute cyanide toxicity and gain a better understanding of the mechanism of action of the hydroxocobalamin in reversal of the toxicity. Swine were intubated, anesthetized, and instrumented with central arterial and venous lines and a pulmonary artery catheter. Animals (n=36) were randomly assigned to one of 3 groups: hydroxocobalamin alone (150 mg/kg), sodium thiosulfate alone (413 mg/kg), or hydroxocobalamin (150 mg/kg)+sodium thiosulfate (413 mg/kg) and monitored for 60 minutes after the start of antidotal infusion. Cyanide was infused until severe hypotension developed, defined as blood pressure 50% of baseline mean arterial pressure. Repeated-measures ANOVA was used to determine statistically significant changes between groups over time. Time to hypotension (25, 28, and 33 minutes), cyanide dose at hypotension (4.7, 5.0, and 5.6 mg/kg), and mean cyanide blood levels (3.2, 3.7, and 3.8 μg/mL) and lactate levels (7, 8.2, 8.3 and mmol/L) were similar. All 12 animals in the sodium thiosulfate group died compared with 2 of 12 in the hydroxocobalamin/sodium thiosulfate group and 1 of 12 in hydroxocobalamin group. No statistically significant differences were detected between the hydroxocobalamin and hydroxocobalamin/sodium thiosulfate groups for carbon monoxide, mean arterial pressure, cyanide levels, or mortality at 60 minutes. Lactate level (2.6 versus 2.1 mmol/L), pH (7.44 versus 7.42), and bicarbonate level (25 versus 26 mEq/L) at 60 minutes were also similar between groups. Sodium thiosulfate failed to reverse cyanide-induced shock in our swine model of severe cyanide toxicity. Further, sodium thiosulfate was not found to be effective when added to hydroxocobalamin in the treatment of cyanide-induced shock. Hydroxocobalamin alone was again found to be effective for severe cyanide toxicity.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium thiosulfate pentahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate solution, 0.01 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.1 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.025 M
Sigma-Aldrich
Sodium thiosulfate solution, 1 M
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP