Skip to Content
Merck
All Photos(4)

Key Documents

217247

Sigma-Aldrich

Sodium thiosulfate pentahydrate

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
grade:
ACS reagent
form:
crystalline powder
crystals
pellets
solubility:
water: soluble(lit.)

grade

ACS reagent

Quality Level

Agency

suitable for EPA 1621
suitable for ISO 25101
suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

Assay

≥99.5%

form

crystalline powder
crystals
pellets

impurities

≤0.002% N compounds
≤0.005% insolubles

pH

6.0-8.4 (25 °C, 5%)

solubility

water: soluble(lit.)

anion traces

S2-: passes test
SO42- and SO32-: ≤0.1%

cation traces

N: ≤0.002%

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium thiosulfate pentahydrate is a hydrated form of sodium thiosulfate. It is used as a sulfur source for the preparation of various organosulfur compounds.

Application

Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.
Sodium thiosulfate pentahydrate can be used as a sulfur source to synthesize:      
  • Thioester derivatives via one-pot two-step reactions with organic halides and aryl anhydrides.      
  • Sulfur nanoparticles using F. benghalensis leaf extract which acts as a reducing and capping agent.      
  • Unsymmetrical heteroaryl thioethers via multicomponent reaction with heteroaryl chlorides and alcohols.

It can also be used in the fabrication of microencapsulated phase change materials for thermal energy storage applications.

Features and Benefits

Na2S2O3 5H2O is a low toxic and odorless reagent that can be easily used in large-scale operations.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fingerprint imaging by scanning electrochemical microscopy.
Zhang M and Girault HH.
Electrochemical Communications, 9(7), 1778-1782 (2007)
Nuclear magnetic resonance in sodium thiosulfate pentahydrate.
Murty CRK and El Saffar ZM.
Acta Crystallographica, 15(6), 536-538 (1962)
Sodium thiosulfate pentahydrate: a redetermination by neutron diffraction.
Lisensky GC and Levy HA.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(6), 1975-1977 (1978)
Fred B. Stocker et al.
Inorganic chemistry, 38(5), 984-991 (2001-10-24)
A new synthetic procedure developed recently in our laboratories has made possible the synthesis of variety of new complexes of CuCN with diamines. Synthesis was effected by adding the ligand to a solution of CuCN in aqueous sodium thiosulfate. This
I A Checheriţă et al.
Chirurgia (Bucharest, Romania : 1990), 108(5), 736-740 (2013-10-26)
In the present article, we discuss the case of 67-year-old female patient diagnosed with inferior limbs calciphylaxis and hemodialyzed since 2006. The clinical manifestations and pathological lab findings are typical for this rare and extremely severe complication in chronic hemodialyzed

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service