Merck
  • Lactate supports cell-autonomous ECM production to sustain metastatic behavior in prostate cancer.
Lactate supports cell-autonomous ECM production to sustain metastatic behavior in prostate cancer.
EMBO reports (2024-06-22)
Luigi Ippolito, Assia Duatti, Marta Iozzo, Giuseppina Comito, Elisa Pardella, Nicla Lorito, Marina Bacci, Erica Pranzini, Alice Santi, Giada Sandrini, Carlo V Catapano, Sergio Serni, Pietro Spatafora, Andrea Morandi, Elisa Giannoni, Paola Chiarugi
ABSTRACT
Extracellular matrix (ECM) is a major component of the tumor environment, promoting the establishment of a pro-invasive behavior. Such environment is supported by both tumor- and stromal-derived metabolites, particularly lactate. In prostate cancer (PCa), cancer-associated fibroblasts (CAFs) are major contributors of secreted lactate, able to impact on metabolic and transcriptional regulation in cancer cells. Here, we describe a mechanism by which CAF-secreted lactate promotes in PCa cells the expression of genes coding for the collagen family. Lactate-exploiting PCa cells rely on increased α-ketoglutarate (α-KG) which activates the α-KG-dependent collagen prolyl-4-hydroxylase (P4HA1) to support collagen hydroxylation. De novo synthetized collagen plays a signaling role by activating discoidin domain receptor 1 (DDR1), supporting stem-like and invasive features of PCa cells. Inhibition of lactate-induced collagen hydroxylation and DDR1 activation reduces the metastatic colonization of PCa cells. Overall, these results provide a new understanding of the link between collagen remodeling/signaling and the nutrient environment exploited by PCa.
Sigma-Aldrich
Anti-P4HA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DDR1 Inhibitor 7rh, ≥98% (HPLC)
Sigma-Aldrich
STAT3 Inhibitor V, Stattic, STAT3 Inhibitor V, Stattic, CAS 19983-44-9, is a cell-permeable inhibitor of STAT3 cellular function that targets the STAT3-SH2 domain and prevents its association with upstream Kinases.