Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chromium(III) potassium sulfate dodecahydrate
Synonym(s):
Chrome alum, Potassium chromium(III) sulfate dodecahydrate, Chromium(III) potassium sulfate dodecahydrate
Linear Formula:
CrK(SO
4
)
2
· 12H
2
O
CAS Number:
7788-99-0
Molecular Weight:
499.40
EC Number:
233-401-6
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
243361
Chromium(III) potassium sulfate dodecahydrate,
ACS reagent, ≥98%
Expand
View Pricing
60152
Chromium(III) potassium sulfate dodecahydrate,
purum p.a., ≥98.5% (RT)
Expand
View Pricing
1.01036
Chromium(III) potassium sulfate dodecahydrate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
05-4560
Chromium(III) potassium sulfate dodecahydrate,
SAJ first grade, ≥97.0%
Expand
View Pricing