1-Ethyl-3-methylimidazolium thiocyanate
Synonym(s):
1-Ethyl-3-methylimidazolium thiocyanate
Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3