Skip to Content
Merck
All Photos(1)

Documents

4.90201

Sigma-Aldrich

1-Ethyl-3-methylimidazolium thiocyanate

for synthesis

Synonym(s):

1-Ethyl-3-methylimidazolium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
MDL number:
UNSPSC Code:
12352107

Quality Level

form

liquid

mp

<-50 °C

density

1.11 g/cm3

InChI

1S/C6H11N2.CHNS/c1-3-8-5-4-7(2)6-8;2-1-3/h4-6H,3H2,1-2H3;3H/q+1;/p-1

InChI key

VASPYXGQVWPGAB-UHFFFAOYSA-M

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service