Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H26N4O4
CAS Number:
Molecular Weight:
434.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
grade
pharmaceutical primary standard
agency
USP
API family
tryptophan
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
format
neat
SMILES string
[n]3(c4c(c(c3)C[C@H](N)C(=O)O)cccc4)C([n]1c2c(c(c1)C[C@H](N)C(=O)O)cccc2)C
InChI
1S/C24H26N4O4/c1-14(27-12-15(10-19(25)23(29)30)17-6-2-4-8-21(17)27)28-13-16(11-20(26)24(31)32)18-7-3-5-9-22(18)28/h2-9,12-14,19-20H,10-11,25-26H2,1H3,(H,29,30)(H,31,32)/t19-,20-/m0/s1
InChI key
DETVQFQGSVEQBH-PMACEKPBSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Tryptophan Related Compound A USP reference standard suitable for use in specified USP compendial tests and assays.
Also used to prepare standard solution during the impurity analysis of Tryptophan by liquid chromatography method coupled to a UV detector.
Also used to prepare standard solution during the impurity analysis of Tryptophan by liquid chromatography method coupled to a UV detector.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Tryptophan Related Compound A
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.