Skip to Content
Merck
All Photos(1)

Key Documents

1235864

USP

Enoxaparin Sodium Molecular Weight Calibrant B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Heparin sodium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

enoxaparin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Enoxaparin Sodium is the sodium salt of a depolymerized heparin. It is obtained by alkaline depolymerization of heparin benzyl ester. Enoxaparin Sodium consists of a complex set of oligosaccharides that have not yet been completely characterized. The majority of the components have a 4-enopyranose uronate structure at the nonreducing end of their chain. Enoxaparin Sodium Molecular Weight Calibrant B reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enoxaparin Sodium Molecular Weight Calibrant B USP reference standard for use in specified quality tests and assays. Also used to plot calibration curves during the determination of molecular weight distribution and weight average molecular weight of Enoxapain Sodium using size exclusion chromatography (SEC).

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enoxaparin Sodium
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 1623-1623 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service