BP561
trans-Clopenthixol hydrochloride
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
trans-Clopenthixol dihydrochloride
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
zuclopenthixol
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
S1c2c(cc(cc2)Cl)\C(=C\CCN4CCN(CC4)CCO)\c3c1cccc3.[Cl-].[Cl-].[H+].[H+]
InChI
1S/C22H25ClN2OS.2ClH/c23-17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)27-22)5-3-9-24-10-12-25(13-11-24)14-15-26;;/h1-2,4-8,16,26H,3,9-15H2;2*1H/b18-5+;;
InChI key
LPWNZMIBFHMYMX-MONHGIHASA-N
General description
Application
Packaging
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service