Skip to Content
Merck
All Photos(1)

Key Documents

BP1210

Ritonavir

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

1,3-Thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48N6O5S2
CAS Number:
Molecular Weight:
720.94
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Quality Level

Agency

BP

API family

ritonavir

form

solid

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 100 mg

manufacturer/tradename

BP

storage condition

dry at room temperature

color

off-white

mp

122-126  °C

solubility

ethanol: soluble
isopropanol: soluble
methanol: soluble

density

1.239 g/cm3 at 20 °C

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CN(C)C(=O)N[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)OCc4[s]cnc4)Cc3ccccc3)Cc2ccccc2)C(C)C

InChI

1S/C37H48N6O5S2/c1-24(2)33(42-36(46)43(5)20-29-22-49-35(40-29)25(3)4)34(45)39-28(16-26-12-8-6-9-13-26)18-32(44)31(17-27-14-10-7-11-15-27)41-37(47)48-21-30-19-38-23-50-30/h6-15,19,22-25,28,31-33,44H,16-18,20-21H2,1-5H3,(H,39,45)(H,41,47)(H,42,46)/t28-,31-,32-,33-/m0/s1

InChI key

NCDNCNXCDXHOMX-XGKFQTDJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ritonavir belongs to the group of protease inhibitors that are widely used in combination with other drugs in the prevention of HIV. Its mode of action involves binding to the active site of the protease enzyme and preventing the further maturation of new viral particles.[1]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ritonavir BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Ritonavir Oral Solution
  • Ritonavir Tablets
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    RP-HPLC method development and validation for simultaneous estimation of Lopinavir and Ritonavir in tablet dosage form
    Bindu MJ, et al.
    International Journal of Science and Engineering Applications, 5(1), 77-77 (2017)
    HPTLC method for simultaneous determination of lopinavir and ritonavir in capsule dosage form
    Sulebhavikar VA, et al.
    Journal of Chemistry, 5(4), 706-712 (2008)

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service