Skip to Content
Merck
All Photos(1)

Key Documents

57677

Frits for Isolation Technologies Precolumn Filter

frit diam. 3 mm

Synonym(s):

Replacement Frit, 3mm, for IT Precolumn Filter, 10 pk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

stainless steel IT frit

packaging

pkg of 10 ea

frit diam.

3 mm

pore size

0.5 μm

compatibility

for use with Isolation Technology Precolumn Filter (57675-U)

Looking for similar products? Visit Product Comparison Guide

General description

The features of Frits for Isolation Technologies Precolumn Filter include:
  • In-line installation.
  • High capacity inlet filter, with 0.5 μm stainless steel frit.
  • Minimizes dead volume and band broadening, to prevent loss of column efficiency while protecting the column.
  • Includes two each of tubing, nut, and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service