Skip to Content
Merck
All Photos(1)

Key Documents

57179

Supelco

Empty SPE Tube (no frits)

volume 12 mL, pk of 20

Synonym(s):

SPE Reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

polypropylene

packaging

pk of 20

technique(s)

solid phase extraction (SPE): suitable

volume

12 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

When used in conjunction with SPE Tube Adapters, empty SPE tubes can be stacked on top of a standard SPE cartridge during processing. The empty SPE cartridge acts as a reseroir to accommodate larger sampling volumes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service