Skip to Content
Merck
All Photos(1)

Key Documents

40327

Supelco

Vinyl acetate solution

certified reference material, 5000 μg/mL in acetonitrile

Synonym(s):

Acetoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

O(C(=O)C)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshifumi Miyazawa et al.
Biotechnology letters, 35(4), 625-630 (2012-12-20)
Acylation of (hydroxyalkyl)phenols with vinyl esters by lipase B from Candida antarctica proceeded smoothly in a highly chemoselective manner, affording their alkyl esters exclusively or at least predominantly. The enzyme therefore discriminates between an alcoholic hydroxyl from a phenolic hydroxyl
María B Blanco et al.
Environmental science & technology, 46(16), 8817-8825 (2012-07-18)
The products formed from the reactions of OH radicals with vinyl acetate and allyl acetate have been studied in a 1080 L quartz-glass chamber in the presence and absence of NO(x) using in situ FTIR spectroscopy to monitor the reactant
Ming-Cheng Lin et al.
Journal of biochemical and molecular toxicology, 25(5), 330-339 (2011-06-02)
Enantiomers of cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate show stereo-specific inhibition for acetylcholinesterase and butyrylcholinesterase. For both inhibition reaction, (2S,4aR,8aS)-cis,cis-decahydro-2-naphthyl-N-n- butylcarbamate is more potent than (2R,4aS,8aR)-cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate. Optically pure (2S,4aR,8aS)-(-)- and (2R,4aS,8aR)-(+)-cis,cis-decahydro-2-naphthols are resolved by the porcine pancreatic lipase-catalyzed acetylation of decahydro-2-naphthols with vinyl acetate. Absolute
B Picquet-Varrault et al.
Environmental science & technology, 44(12), 4615-4621 (2010-05-25)
Vinyl acetate is widely used in industry. It has been classified as a high-production volume (HPV) chemical in the United States. To evaluate its impact on the environment and air quality, its atmospheric reactivity toward the three main tropospheric oxidants
I Lara-Mayorga et al.
Environmental technology, 31(1), 1-6 (2010-03-18)
In a previous paper, the authors showed that a slight aeration of a methanogenic reactor treating wastewater from the manufacture of polymeric resins could improve its performance, by increasing or allowing the removal of some of its contaminants, including vinyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service