29742-U
Tenax® GR Thermal Desorption Tube
stainless steel, O.D. x I.D. x L 1/4 in. × 3 1/2, preconditioned, pkg of 10 ea
Synonym(s):
Tenax® GR
About This Item
Recommended Products
Product Name
Tenax® GR, 60/80 mesh, O.D. × L 1/4 in. × 3 1/2, stainless steel TD tube, sealed with Brass Endcaps, preconditioned, pkg of 10 ea
material
stainless steel TD tube
feature
preconditioned
packaging
pkg of 10 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
technique(s)
active air sampling: suitable
O.D. × L
1/4 in. × 3 1/2
matrix
Tenax GR graphitized porous polymer
particle size
60/80 mesh
suitability
suitable for (extends range of Tenax TA)
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service