ZRB2785
Anti-PSAT1 Antibody, clone 1C7 ZooMAb® Rabbit Monoclonal
![Enhanced Validation antibodies are tested to ensure reproducibility, specificity, and performance using our enhanced validation strategies enhanced validation](/static/enhanced_validation_badge.png)
recombinant, expressed in HEK 293 cells
Synonym(s):
EC:2.6.1.52, Phosphohydroxythreonine aminotransferase, PSAT, Phosphoserine aminotransferase
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
recombinant monoclonal
description
clone 1C7
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 40.4 kDa
observed mol wt ~39 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
Internal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... PSAT1(29968)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in A549 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PSAT1 in A549 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PSAT1 in Human brain tissue lysate.
Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected PSAT1 in human kidney tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PSAT1 in human kidney tissue sections.
Western Blot using lysates from wild-type(WT) and PSAT1 knockout cells: A 1:5,000 dilution from a representative lot detected PSAT1 in lysates from NSCLC cells and NSCLC cells with PSAT1 knock-down and transfected to re-express WT PSAT1, but not in PSAT1 knock-down or mock transfected cells. (Data courtesy Dr. Brian Clem, University of Louisville)..
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service