ZMS1180
Anti-LAG3 Antibody, clone 4-10-C9 ZooMAb® Mouse Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Anti-LAG3 Antibody, clone 4-10-C9 ZooMAb® Mouse Monoclonal, Activation-induced cytidine deaminase-linked autoimmunity protein, Aida, CD223, LAG-3, Lymphocyte activation gene 3 protein
About This Item
Recommended Products
biological source
mouse
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
4-10-C9, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 56.98 kDa
observed mol wt ~80 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG2aκ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
mouse ... Lag3(16768)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting with recombinant fragment corresponding to the extracellular domain of Mouse LAG3.
Western Blotting (WB) Analysis: A1:1,000 dilution of this antibody detected recombinant fragment corresponding to the extracellular domain of Mouse LAG3 fused to Fc portion of mouse IgG2a.
Tested Applications
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected LAG3 in Mouse splenocytes treated with Concanavalin A (2 g/mL; 3 days).
Affinity Binding Assay:: A representative lot of this antibody bound recombinant fragment corresponding to the extracellular domain of Mouse LAG3 with a KD of 2.8 x 10-6 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service