Z744240
Wilmad® 3.2 mm Bruker® MAS Rotor Set
includes 2 Vespel™ caps and bottom plugs
Synonym(s):
Solid State NMR, MAS NMR,
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pack of 1 ea (2 caps and plugs)
manufacturer/tradename
Wilmad WP-501-3180-SET1
General description
Manufactured from the highest purity Zirconia, MAS Rotors provide the NMR spectroscopist with the ultimate alternative for analysis of solid samples; the need to solvate is eliminated. They are available for most present solid state NMR spectrometers. MAS Rotors are carefully examined for material irregularities by optical methods before and after the precision machining process. Without over-spinning, each rotor is spin tested to only the highest specified spinning speed. Overspin testing, a common practice with some rotor brands, can actually damage the rotor prior to use. High precision (which becomes a necessity for proper spinning performance) is maintained in the manufacture of the end caps. Most caps are fitted with O-rings for better sealing; some have axial holes for venting. Advantages of Zirconia Rotors Most homogeneous ceramic material structure. Zirconia Rotor Body has higher strength (1,000 MPa, greater than Si3N4). Highest spinning speed, high precision superior finish (0.05 µ mirror gloss). Some caps have a uniquely designed detachable O-ring for use when a gas-tight environment is required for the sample.
Application
NMR, MAS NMR
Features and Benefits
- Homogeneous ceramic material structure.
- Zirconia Rotor Body strength 1,000 MPa
- Finish 0.05 µ mirror gloss
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Vespel is a trademark of E. I. du Pont de Nemours and Company
Wilmad is a registered trademark of SP Industries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service