Key Documents
Z740156
LifeSep magnetic separation stand
15SX, for 15 mL tubes
Synonym(s):
magnetic separation equipment
About This Item
UNSPSC Code:
41100000
NACRES:
NA.56
feature
for 15 mL tubes
manufacturer/tradename
Dexter Magnetic Technologies 2501023
Related Categories
General description
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Open-faced design for clear visibility
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
  • Durable and easy to use
Choose from one of the most recent versions:
Certificates of Analysis (COA)