Z711020
Media bottles, wide mouth, Lab 45, accessory
red ETFE pour ring
Synonym(s):
240760
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
red ETFE pour ring
manufacturer/tradename
WHEATON®
Features and Benefits
- For high temperature applications, use the PBT (polybutylene terephthalate) screw caps and the ETFE (ethylene tetrafluoroethylene) pour lips which can withstand temperatures up to 200ºC up to 2 hours
- Autoclavable
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)