Z688169
HandEvac handheld aspirator
complete operator set
Synonym(s):
Handheld Aspirator, Handheld Vacuum Aspirator, Portable Aspirator
All Photos(1)
About This Item
UNSPSC Code:
41105100
NACRES:
NB.22
Recommended Products
manufacturer/tradename
Argos 1305032
General description
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes!
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Includes: EV526, EV501, EV520 and EV503
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Includes: EV526, EV501, EV520 and EV503
Choose from one of the most recent versions:
Certificates of Analysis (COA)