Skip to Content
Merck
All Photos(4)

Key Documents

WH0009588M1

Sigma-Aldrich

Monoclonal Anti-PRDX6 antibody produced in mouse

clone 3A10-2A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-1Cys, Anti-AOP2, Anti-KIAA0106, Anti-MGC46173, Anti-NSGPx, Anti-PRX, Anti-aiPLA2, Anti-p29, Anti-peroxiredoxin 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A10-2A11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDX6(9588)

Related Categories

General description

Peroxiredoxin 6 (Prdx6) is a non-seleno peroxidase enzyme, encoded by the gene mapped to human chromosome 1q25.1. Prdx6 belongs to thiol-specific antioxidant protein family. It is highly expressed in lung, alveolar epithelial type II cells and alveolar macrophages. The protein is characterized with a single conserved cysteine residue and is called 1-Cys Prdx. Prdx6 contains 224 amino acids.

Immunogen

PRDX6 (AAH35857.1, 1 a.a. ~ 224 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPGGLLLGDVAPNFEANTTVGRIRFHDFLGDSWGILFSHPRDFTPVCTTELGRAAKLAPEFAKRNVKLIALSIDSVEDHLAWSKDINAYNCEEPTEKLPFPIIDDRNRELAILLGMLDPAEKDEKGMPVTARVVFVFGPDKKLKLSILYPATTGRNFDEILRVVISLQLTAEKRVATPVDWKDGDSVMVLPTIPEEEAKKLFPKGVFTKELPSGKKYLRYTPQP

Biochem/physiol Actions

Peroxiredoxin 6 (Prdx6) is a bifunctional enzyme, carrying out both phospholipase A2 (PLA2) activity and glutathione (GSH) peroxidase function. The encoded protein aids in redox regulation of the cell and is implicated in reduction of H2O2 and short chain organic, fatty acid, and phospholipid hydroperoxides. It might also have a crucial role in the regulation of phospholipid turnover and in antioxidant defense. Prdx6 is highly expressed at leading edges of wounds and aids in wound healing. Deficiency of the enzyme activity is observed in cataracts of the lens. Overexpression of Prdx6 leads to several degenerative CNS (central nervous system) disorders such as Alzheimer′s, Parkinson′s, Pick′s, and Crutzfeld-Jakob diseases and schizophrenia. In addition, elevated expression of the enzyme is also observed in various neoplastic diseases including malignant mesothelioma, bronchogenic squamous cell carcinoma and cancer of the gingivo-buccal area, bladder and breast.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxiredoxin 6 as an antioxidant enzyme: protection of lung alveolar epithelial type II cells from H2O2-induced oxidative stress.
Wang Y
Journal of Cellular Biochemistry, 104(4) , 1274-1285 (2008)
Daniel Martins-de-Souza et al.
BMC psychiatry, 9, 17-17 (2009-05-02)
Schizophrenia is likely to be a consequence of DNA alterations that, together with environmental factors, will lead to protein expression differences and the ultimate establishment of the illness. The superior temporal gyrus is implicated in schizophrenia and executes functions such
Proteome analysis of schizophrenia patients Wernicke's area reveals an energy metabolism dysregulation.
Matins-de-Souza, et.al
BMC Psychiatry, 9, 17-17 (2009)
Peroxiredoxin 6: a bifunctional enzyme with glutathione peroxidase and phospholipase A2 activities.
Fisher AB
Antioxidants & Redox Signaling, 15(3) , 831-844 (2011)
Rimpy Kaur Chowhan et al.
Scientific reports, 10(1), 17416-17416 (2020-10-17)
Peroxiredoxin 6 (Prdx6) is a ubiquitously expressed antioxidant non-selenium glutathione peroxidase that is known to play a major role in various physiological and pathological processes. It belongs to the family of peroxidases (referred to as Peroxiredoxins, Prdx's) that work independently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service