Skip to Content
Merck

Skip To

UPS3

Universal Proteomics Standard 3

Protein Mass Spectrometry Calibration Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

ready-to-use solution

quality

Protein Mass Spectrometry Calibration Standard

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

General description

Universal Proteomics Standard 3 (UPS3) is a mixture of 8 non-human proteins at 4 orders of magnitude. It is intended to be useful as a quantitative reference standard for samples containing human proteins and analyzed by mass spectrometry. The proteins were screened for limited human homology in silico, and tested by peptide mapping to ensure little or no interference with human samples.

Protein Sequence (FASTA)

Application

Proteomic Quantification

Features and Benefits

In an effort to facilitate standardized, optimized, and shareable data/protocols for analysis of human protein samples, we have designed a new standard:
  • Affordable, for routine use
  • No peptide homology with human samples
  • Minimized carryover between runs

Other Notes

EMS0004 (SOLu-Trypsin) and MSKT0002 (SOLu-Trypsin Rapid Digestion Kit)
This product is comprised of the following: Each vial contains eight non-human, lyophilized proteins ranging from 100 picomoles to 0.1 picomoles. Total protein content is ~8 μg as determined by UV.

Disclaimer

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UPS2MSST0059UPS1
form

ready-to-use solution

form

ready-to-use solution

form

lyophilized powder

form

ready-to-use solution

technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

-

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

quality

Protein Mass Spectrometry Calibration Standard

quality

Protein Mass Spectrometry Calibration Standard

quality

-

quality

Protein Mass Spectrometry Calibration Standard


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters