TTR005
Gram Stain TISSUE-TROL™ Control Slides
mouse lung tissue containing Staphylococcus aureus and Escherichia coli
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116121
NACRES:
NA.47
Recommended Products
biological source
mouse lung (tissue containing Staphylococcus aureus and Escherichi coli)
application(s)
hematology
histology
storage temp.
room temp
Application
Histology slides with paraffin embedded section of mouse lung tissue containing Staphylococcus aureus and Escherichia coli. Suitable for use with Gram Stain Procedure No. HT90.
Components
Box contains 24 unstained slides plus one reference slide stained using Gram Stain Procedure No. HT90.
Legal Information
TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service