T6649
Microcentrifuge tubes with attached lid
capacity 1.5 mL
Synonym(s):
Microcentrifuge Tubes, Tubes with Attached Lid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cap (Attached lid)
clear
polypropylene
sterility
non-sterile
packaging
case of 1,000 ("Case of 1000)
pack of 500
H × O.D.
38 mm × 11 mm
capacity
1.5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Microcentrifuge tubes with attached lids have been used:
- in thecollection and thawing of bovine sperm samples
- in glucose sensor severeacute respiratory syndrome coronavirus 2 (SARS-CoV-2) assay andglucometer-based SARS-CoV 2 assay
- in the preparation of plasmid DNAsolutions
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Data in brief, 38, 107278-107278 (2021-08-18)
We present supplementary data for the published article, "Hitting the diagnostic sweet spot: Point-of-care SARS-CoV-2 salivary antigen testing with an off-the-shelf glucometer" [1]. The assay described is designed to be performed at home or in a clinic without expensive instrumentation
Methods and protocols, 4(4) (2021-10-27)
Sperm binding within the oviductal sperm reservoir plays an important role for reproductive success by enabling sperm survival and maintaining fertilizing capacity. To date, numerous in vitro technologies have been established to measure sperm binding capacity to cultured oviductal cells
Bio-protocol, 7(15), e2432-e2432 (2017-08-05)
Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR) and CRISPR-associated (Cas) systems function as an adaptive immune system in bacteria and archaea for defense against invading viruses and plasmids (Barrangou and Marraffini, 2014). The effector nucleases from some class 2 CRISPR-Cas
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service