Skip to Content
Merck
All Photos(1)

Key Documents

T0449

Sigma-Aldrich

Triethanolamine Buffer solution

Synonym(s):

TEA Buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.21

form

liquid

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Application


  • Analysis of Cationic Vitamins in Cell Culture Medium Samples by Capillary Zone Electrophoresis.: While the study focuses on the analysis of cationic vitamins, the use of Triethanolamine buffer in electrophoresis provides an insight into its application in bioanalytical chemistry, useful for biochemists working with cell cultures and analytical methods (van der Burg D et al., 2022).

  • Fucoidan-based hydrogels particles as versatile carriers for diabetes treatment strategies.: This article discusses the development of fucoidan-based hydrogels, where Triethanolamine may play a role in stabilizing or modifying biochemical properties, essential for biochemists researching drug delivery systems (Reys LL et al., 2022).

  • Cationic Side Chain Identity Directs the Hydrophobically Driven Self-Assembly of Amphiphilic ß-Peptides in Aqueous Solution.: The study provides information on the interaction of cationic side chains with buffers like Triethanolamine, which is crucial for biochemists studying peptide self-assembly and molecular interactions (Wang C et al., 2021).

  • Electrochemistry and Electrochemiluminescence of Coumarin Derivative Microrods: Mechanism Insights.: Examines the electrochemical behaviors where Triethanolamine buffer may be involved, offering valuable insights for biochemists interested in electrochemical analysis and sensor technology (Yan M et al., 2021).

Preparation Note

Contains triethanolamine, 0.2 mol/L, magnesium ions and EDTA, pH 8.0 at 25 °C. Less than 0.1% sodium azide added as a preservative.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Young Lee et al.
Journal of analytical toxicology, 42(5), 293-299 (2018-04-05)
A sensitive method for the purification and determination of two protein adducts, organophosphorus (OP)-BChE and OP-albumin adducts, in a single sample using a simultaneous sample preparation method was developed and validated using liquid chromatography-tandem mass spectrometry. First, we isolated O-ethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service